Learn More
Thomas Scientific Gas Permeable Clear Heat Seal; clear heat sealing film, with 3 mm slits for gas transfer, 100 sheets (125 x 78 mm)
SDP
Catalog No. NC0735423
Encompass
Peelable heat sealing film, optically clear; suitable for qPCR and optical applications Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. Our Clear Heat Seal is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates. Samples can also be accessed by pre-piercing with a blade, needle or our Pierce Plate (4ti-0398). The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays. The Clear Heat Seal forms a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurised heated lid). This seal demonstrates moderate solvent resistance and can be utilised for short term compound storage at room temperat
Catalog No. NC0735423 Supplier Thomas Scientific Supplier No. 1149T29
Peelable heat sealing film, optically clear; suitable for qPCR and optical applications Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. Our Clear Heat Seal is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates. Samples can also be accessed by pre-piercing with a blade, needle or our Pierce Plate (4ti-0398). The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays. The Clear Heat Seal forms a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurised heated lid). This seal demonstrates moderate solvent resistance and can be utilised for short term compound storage at room temperat