Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Marketlab Clear Image Singles
Description
- Individual patient packet designed for reduced cross-contamination risks
- Built-in dispensing spout helps clinician control the flow of gel
- Manufactured utilizing a proprietary method to ensure minimal air bubbles for improved image clarity
- Water-based, water-soluble formula
- Clear and nonstaining
- Contains no irritating dyes, perfumes, parabens, or surfactants
- Does not contain oils, alcohol, silicones, or fragrance
- Bacteriostatic
- Nonsterile, for external use only (not for use in electromedical procedures)
Specifications
Specifications
| For Use With (Application) | High Viscosity Gel |
| Quantity | 400/Pk. |
| Packaging | Dispensing Carton |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?