Learn More
Alkali Scientific Clear Immunoplate PS 96 Well P
SDP
Catalog No. 503028491
Encompass_Preferred
ASI96-Well Solid Plates Clear polystyrene surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assaysAlphanumerically labeled rows and columns Multibinding Suitable specifically for adsorbing hydrophilic molecules (Antigen ELISA) Maxibinding Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions (Recommended for most ELISA experiments) Medibinding Surface adsorbs proteins and molecules that consists of interrmediate hydrophobic and hydrophilic properties (Antigen/Antibody ELISA More hydrophbic compared to Maxibinding Plates) Unibinding Suitable for adsorption of hydrophobic molecules (Antigen ELISA) Plate type (96well per plate) Lid not included 3 type bottom Flat / Round / Flat-Small Developed for immunoassay(ELISA) and general bin
Catalog No. 50-302-8491 Supplier Alkali Scientific Supplier No. MPFSUNI
Add to Cart
Add to Cart
ASI96-Well Solid Plates Clear polystyrene surface chemistry for optimal binding is hydrophilic and ideal for protein research and antibody sandwich assaysAlphanumerically labeled rows and columns Multibinding Suitable specifically for adsorbing hydrophilic molecules (Antigen ELISA) Maxibinding Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions (Recommended for most ELISA experiments) Medibinding Surface adsorbs proteins and molecules that consists of interrmediate hydrophobic and hydrophilic properties (Antigen/Antibody ELISA More hydrophbic compared to Maxibinding Plates) Unibinding Suitable for adsorption of hydrophobic molecules (Antigen ELISA) Plate type (96well per plate) Lid not included 3 type bottom Flat / Round / Flat-Small Developed for immunoassay(ELISA) and general bin