Learn More
Electron Microscopy Sciences Clear-Mount with Tris Buffer 250 ML
SDP
Catalog No. 5033566
Encompass_Preferred
Clear-Mount is a water based mounting medium designed for the permanent mounting of hydrated tissues, which may be damaged with organic solvents. Such samples include cell smears, with peroxidase and alkaline phosphatase chromogens.
This mounting medium preserves Fast Red, Aminoethylycarbazole (AEC), NBT/BCIP, INT/BCIP chromogens and is also compatible with counter-stain such as Hematoxylin and Nuclear Fast Red. It is also suitable for chromogens like DAB and DAB with Nickel and Cobalt. It is not compatible with H and E staining.
Application: Mounting of Immunohisto slides.
Catalog No. 50-335-66 Supplier Electron Microscopy Sciences Supplier No. 1798515
Clear-Mount is a water based mounting medium designed for the permanent mounting of hydrated tissues, which may be damaged with organic solvents. Such samples include cell smears, with peroxidase and alkaline phosphatase chromogens.
This mounting medium preserves Fast Red, Aminoethylycarbazole (AEC), NBT/BCIP, INT/BCIP chromogens and is also compatible with counter-stain such as Hematoxylin and Nuclear Fast Red. It is also suitable for chromogens like DAB and DAB with Nickel and Cobalt. It is not compatible with H and E staining.
Application: Mounting of Immunohisto slides.