Learn More
LIFECANVAS TECHNOLOGIES INC Clear+ Passive Clearing Kit, 8 sample kit
SDP
Catalog No. NC3593548
Encompass
The 8 sample kit comes with 250 mL SHIELD kit (SHIELD Epoxy, Buffer, and ON solutions), 250 mL Clear+ Delipidation Buffer, and 250 mL EasyIndex with Refractive Index 1.52. Our reagents preserve fluorescent proteins, reduce autofluorescence, and optimally prepare samples for immunostaining, 3D imaging, and analysis. Click
Catalog No. NC3593548 Supplier LIFECANVAS TECHNOLOGIES INC Supplier No. CPCK2501.52
The 8 sample kit comes with 250 mL SHIELD kit (SHIELD Epoxy, Buffer, and ON solutions), 250 mL Clear+ Delipidation Buffer, and 250 mL EasyIndex with Refractive Index 1.52. Our reagents preserve fluorescent proteins, reduce autofluorescence, and optimally prepare samples for immunostaining, 3D imaging, and analysis. Click