Learn More
Omnimed Inc Clear PETG Glove Box Holder
SDP
Catalog No. NC3956475
Encompass_Preferred
Omnimed plastic glove box holder are made of Eco friendly PETG plastic that is both durable and easy to clean. These holders will withstand years of use and still look new. They are ideal for use in medical, lab and food prep functions or any area where cross contamination is an issue or protective gloves are needed. These holders accommodate one glove box, are simple to load and can be mounted vertically or horizontally on a variety of surfaces. All single glove boxes fit this model.
Catalog No. NC3956475 Supplier Omnimed Inc Supplier No. 305360
Omnimed plastic glove box holder are made of Eco friendly PETG plastic that is both durable and easy to clean. These holders will withstand years of use and still look new. They are ideal for use in medical, lab and food prep functions or any area where cross contamination is an issue or protective gloves are needed. These holders accommodate one glove box, are simple to load and can be mounted vertically or horizontally on a variety of surfaces. All single glove boxes fit this model.