Learn More
Sklar Instruments Clear Plastic Tray, Sklar, Large. 11in. x 13 in. x 6 in. Deep. Three piece set includes tray, strainer and lid. For Storage/Transportation Only. PK/3.
SDP
Catalog No. 501437183 Shop All Sklar Instruments Products
Encompass_Preferred
Clear Plastic Tray, Sklar, Large. 11 in. x 13 in. x 6 in. Deep. Three piece set includes tray, strainer and lid. Clear Plastic. For Storage/Transportation Only. Sklar's Clear Plastic Trays are non-sterile containers and used for storage and transportation of instruments. They are not intended for use with sterilization or cleaning products. The trays are available in a variety of sizes.
This item is not returnable. View return policy
Catalog No. 50-143-7183 Supplier Sklar Instruments Supplier No. 103060
Clear Plastic Tray, Sklar, Large. 11 in. x 13 in. x 6 in. Deep. Three piece set includes tray, strainer and lid. Clear Plastic. For Storage/Transportation Only. Sklar's Clear Plastic Trays are non-sterile containers and used for storage and transportation of instruments. They are not intended for use with sterilization or cleaning products. The trays are available in a variety of sizes.