Learn More
3M™ Clear Polycarbonate Faceshield
Catalog No. 19070902 Shop All 3M Products
Change view
Click to view available options
Clear
Catalog No.
19-070-902 Clear
1 options
Catalog No. 19-070-902 Supplier 3M™ Supplier No. 7000002337
Add to Cart
Add to Cart
3M™ Clear Polycarbonate Faceshield helps provide high impact face protection.
  • Flat stock.
  • Designed to help provide face protection to workers in metalworking, utilities, forestry and many other industries.
  • Meets the high impact requirements of ANSI Z87+
  • For use with 3M™ Chin Protector CP8 only and any 3M™ Headgear.
  • Mounts quickly to any 3M headgear without tools, snaps or cams.
TRUSTED_SUSTAINABILITY
Specifications
Model 82543-00000