Learn More
Analytical Sales & Services Clear PP Domed Caps for 600µL PP Tubes, Strips of 12
Catalog No. 5000805127
Encompass
Advantage Clear Polypropylene Domed Cap Strips\n• Use for 600µL Polypropylene Tubes\n• 12 Caps/Strip\n\nQty/Pkg: 125
Catalog No. 50-008-05127 Supplier Analytical Sales & Services Supplier No. 96840
Advantage Clear Polypropylene Domed Cap Strips\n• Use for 600µL Polypropylene Tubes\n• 12 Caps/Strip\n\nQty/Pkg: 125