Learn More
Cole-Parmer Cole-Parmer Clear Precleaned EPA vials 60 mL, 72/cs
Supplier:  Cole-Parmer 9953525
Encompass_Preferred
These glass vials are precleaned to meet strict EPA requirements. Vials with certificate of analysis meet EPA criteria for more than 60 volatile organic analyses (VOA). All vials include a polypropylene cap with a 1/8 inch PTFE-faced silicone septum.
Catalog No. NC1859662