Learn More
MSC Dymon Glass Cleaner, Aerosol, 19 oz, Ammonia-Free
Catalog No. 502837284
Encompass
This glass cleaner is formulated to effectively remove common contaminants such as grease, fingerprints, oil, smudges, soils, and streaks from surfaces. Its ammonia-free, foaming aerosol formula clings to vertical surfaces and dries clear, leaving no spots, streaks, or film behind, even in varying temperatures.
  • Removes grease, fingerprints, oil, smudges, soils, and streaks
  • Overspray dries clear
  • Leaves no spots, streaks, or film behind at either warm or cold temperatures
  • Leaves no residue
  • Foaming formula clings to vertical surfaces
  • Ammonia-free formula
  • Fresh, clean fragrance
Catalog No. 50-283-7284 Supplier MSC Supplier No. 66554635
This glass cleaner is formulated to effectively remove common contaminants such as grease, fingerprints, oil, smudges, soils, and streaks from surfaces. Its ammonia-free, foaming aerosol formula clings to vertical surfaces and dries clear, leaving no spots, streaks, or film behind, even in varying temperatures.
  • Removes grease, fingerprints, oil, smudges, soils, and streaks
  • Overspray dries clear
  • Leaves no spots, streaks, or film behind at either warm or cold temperatures
  • Leaves no residue
  • Foaming formula clings to vertical surfaces
  • Ammonia-free formula
  • Fresh, clean fragrance