Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
GSA_VAGREENER_CHOICE

Catalog No. 1424577
Change view
Click to view available options
Surface Treatment:
Immulon 1 B
Immulon 2 HB
MaxiSorp
PolySorp
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
14-245-77 Immulon 1 B
14-245-79 Immulon 2 HB
12-565-137 MaxiSorp
12-565-139 PolySorp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Each lot is functionally tested for IgG binding.
Catalog No. 14-245-77 Supplier Thermo Scientific™ Supplier No. 3555
Add to Cart
Add to Cart
Each lot is functionally tested for IgG binding.

Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces

  • PolySorp and Immulon 1B for adsorption of hydrophobic molecules
  • Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays

Ordering Information:

Please note that availability may vary by country.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophobic
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 280 μL
No. of Wells 96
Volume (Metric) Working 180 μL
Sterility Non-sterile
Surface Treatment Immulon 1 B
Well Shape Round
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Lipids
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.