Learn More
Clear Safety Goggles
Catalog No. 12888303 Shop All Fisherbrand Products
Change view
Click to view available options
Safety Goggles; Color: Clear; Fog and scratch resistant
Catalog No.
12-888-303 Safety Goggles; Color: Clear; Fog and scratch resistant
1 options
This item is not returnable. View return policy
Catalog No. 12-888-303
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Safety goggles providing eye protection
  • Indirect ventilation
  • Anti-fog
  • Anti-scratch
  • Non-sterilized
  • PC 2.0 mm lens (EN 166 optical class 1)
  • ANSI Z87+ D3 and D4
  • High speed impact resistant in extreme temperatures (EN 166 BT)
  • Fits over prescription glasses
Specifications
Certifications/Compliance ANSI Z87.1 D3D4, EN166 1 BT
Color Clear
Lens Tint Clear
Product Type Safety goggles
Frame Color Clear
Size 17 x 8 cm
Disposable Reusable
Frame Material Fog and scratch resistant