Learn More
Fisherbrand™ Clear Safety Goggles
Catalog No. 19181512 Shop All Fisherbrand Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Catalog No. Lens Coating
19-181-512 Anti-fog, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-181-512
Add to Cart
Add to Cart
This item is not returnable. View return policy
Comfortable eye protection
  • Ventilated
  • Anti-fog
  • Anti-scratch
  • ANSI Z87+1B  and D4
Compliance
Meets ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4 standards.
Specifications
Certifications/Compliance ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Ventilated Safety Goggles
For Use With (Application) Eye protection
Frame Color Clear