Learn More
Grainger Clear Sausage Barrel Kit 20 Oz
Catalog No. 502748766
Encompass_Preferred
The Milwaukee Caulk Gun Barrel Sausage Pack is a caulk gun accessory designed to dispense caulk, adhesives and sealants from sausage packs. It features a steel barrel with a smooth rod that dispenses material in a continuous flow without dripping. This barrel attaches to all Milwaukee caulk guns and accepts 10 oz and 20 oz sausage packs. It has a maximum pressure of 1100 psi. The barrel is 8 inches long and made from carbon steel for durability. This allows easy dispensing from sausage packs without the need to cut open packs.
Catalog No. 50-274-8766 Supplier Grainger Supplier No. 14P351
The Milwaukee Caulk Gun Barrel Sausage Pack is a caulk gun accessory designed to dispense caulk, adhesives and sealants from sausage packs. It features a steel barrel with a smooth rod that dispenses material in a continuous flow without dripping. This barrel attaches to all Milwaukee caulk guns and accepts 10 oz and 20 oz sausage packs. It has a maximum pressure of 1100 psi. The barrel is 8 inches long and made from carbon steel for durability. This allows easy dispensing from sausage packs without the need to cut open packs.