Learn More
CELLTREAT Scientific Products Clear Sealing Film, Pressure Activated, Non-sterile
SDP
Catalog No. 5000144350
Encompass_Preferred
Clear Sealing Film is optically clear with low auto-fluorescence, ideal for qPCR applicationsPressure-activated adhesive is non-tacky and creates a strong bond when applied with a sealing paddle or rollerNon-activated portions remain non-tacky, making this film suitable for dry samples
Catalog No. 50-001-44350 Supplier CELLTREAT Scientific Products Supplier No. 229134
Clear Sealing Film is optically clear with low auto-fluorescence, ideal for qPCR applicationsPressure-activated adhesive is non-tacky and creates a strong bond when applied with a sealing paddle or rollerNon-activated portions remain non-tacky, making this film suitable for dry samples