Learn More
Mitegen HD Clear Sealing Tape, 3-Inch Wide, 55-Yard Roll
SDP
Catalog No. 501941001
Encompass_Preferred
The HD Clear Sealing Tape is a highly effective sealant specifically designed for optimizing sitting drop crystallization experiments. This extra-wide, 3-inch tape (7.62 cm) is supplied as a generous 55-yard roll, providing ample material for numerous crystallization plates. Its exceptional clarity ensures unobstructed visualization of growing crystals, which is crucial for monitoring progress and assessing crystal quality without removal. As an ideal sealant, it effectively prevents evaporation and contamination within crystallization wells, maintaining stable conditions essential for successful crystal growth. The tape's robust adhesive ensures a secure, long-lasting seal, providing peace of mind during extended incubation periods.
Catalog No. 501941001 Supplier Mitegen Supplier No. MCTP101M
The HD Clear Sealing Tape is a highly effective sealant specifically designed for optimizing sitting drop crystallization experiments. This extra-wide, 3-inch tape (7.62 cm) is supplied as a generous 55-yard roll, providing ample material for numerous crystallization plates. Its exceptional clarity ensures unobstructed visualization of growing crystals, which is crucial for monitoring progress and assessing crystal quality without removal. As an ideal sealant, it effectively prevents evaporation and contamination within crystallization wells, maintaining stable conditions essential for successful crystal growth. The tape's robust adhesive ensures a secure, long-lasting seal, providing peace of mind during extended incubation periods.