Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Clear Serum Bottle, Crimp Top

Catalog No. 111016329 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
30 mL
50 mL
100 mL
No. per Pack:
144/Pk.
272/Pk.
288/Pk.
36/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) No. per Pack
11-101-6329 50 mL 272/Pk.
11-101-0959 100 mL 36/Pk.
11-100-9449 100 mL 144/Pk.
11-100-9575 30 mL 288/Pk.
11-100-9973 2.5 mL 288/Pk.
11-101-1544 30 mL 36/Pk.
11-102-0917 50 mL 36/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11-101-6329 Supplier MilliporeSigma™ Supelco™ Supplier No. 33109U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constructed of borosilicate glass, closures are not included

TRUSTED_SUSTAINABILITY

Specifications

Description No Closure Included
Diameter (Metric) Opening 20 mm
Color Clear
Diameter (Metric) Outer 43 mm
Material Glass
Volume (Metric) 50 mL
No. per Pack 272/Pk.
Closure Type Crimp Cap
Autoclavable Autoclavable
Height (Metric) 73 mm
Product Type Serum Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.