Learn More
Electron Microscopy Sciences Clear Silicone Rubber Kit 1 GL
SDP
Catalog No. 5031094
Encompass_Preferred
This silicone cures at room temperature with no shrinkage and offers a unique water clear see through rubber with excellent tear strength, chemical, and heat resistance.
Allow the mold to cure overnight (at least 16 hours) at room temperature (77 degree F/25 degree C) before demolding. Do not cure rubber where temperature is less than 65 degree F/18 degree C. Post curing the mold will aid in quickly attaining maximum physical and performance properties. After curing at room temperature, expose the rubber to 80 degree C for 2 hours and 100 degree C for 2 hours. Allow mold to cool to room temperature before using.
NOTE: This clear silicone will cause rubber to yellow when applying post curing. Clear silicone comes as a kit of 1 gallon (9 lb) silicone A and 1 pint (.9lb) curing agent B.
Catalog No. 50-310-94 Supplier Electron Microscopy Sciences Supplier No. 24234C
This silicone cures at room temperature with no shrinkage and offers a unique water clear see through rubber with excellent tear strength, chemical, and heat resistance.
Allow the mold to cure overnight (at least 16 hours) at room temperature (77 degree F/25 degree C) before demolding. Do not cure rubber where temperature is less than 65 degree F/18 degree C. Post curing the mold will aid in quickly attaining maximum physical and performance properties. After curing at room temperature, expose the rubber to 80 degree C for 2 hours and 100 degree C for 2 hours. Allow mold to cool to room temperature before using.
NOTE: This clear silicone will cause rubber to yellow when applying post curing. Clear silicone comes as a kit of 1 gallon (9 lb) silicone A and 1 pint (.9lb) curing agent B.