Learn More
Bellco Glass, Inc. Clear Snap Cap W/PTFE/Sil,11mm
SDP
Catalog No. 501945401
Encompass_Preferred
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe
Catalog No. 50-194-5401 Supplier Bellco Glass, Inc. Supplier No. 207003200
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe