Learn More
Alkali Scientific ASI™ Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps, 32 oz., 12 Case
SDP
Catalog No. 503028432
Encompass_Preferred
Clear, Type III soda-lime glass ideal for general, small to large volume storage
Catalog No. 50-302-8432 Supplier Alkali Scientific Supplier No. WMB032
Clear, Type III soda-lime glass ideal for general, small to large volume storage