Learn More
Molecular Dimensions CLEAR STRATEGY SCREEN 1 HT-96
SDP
Catalog No. NC0090593
Encompass
Protein Crystallography. Clear Strategy Screen I HT-96 (96 x 1.1mL): allows users to easily interpret results & plan optimization. In the HT-96 format, the Clear Strategy I reagents are premixed at 4 pH values (pH's 5.5, 6.5, 7.5 and 8.5).
Catalog No. NC0090593 Supplier Molecular Dimensions Supplier No. MD131
Protein Crystallography. Clear Strategy Screen I HT-96 (96 x 1.1mL): allows users to easily interpret results & plan optimization. In the HT-96 format, the Clear Strategy I reagents are premixed at 4 pH values (pH's 5.5, 6.5, 7.5 and 8.5).