Learn More
Med Vet International Clear Thera-Gel 1/8” Thick Sheet, 4in x 4in, J0940S, Each
Catalog No. 5000581362
Encompass_Preferred
With self-adhesive back. Cut to fit to protect bony prominences and sensitive areas. Contains mineral oil to help soften and protect delicate skin. Washable with soap and water. Latex-free, 4" x 4".
Catalog No. 50-005-81362 Supplier Med Vet International Supplier No. JOR940S
With self-adhesive back. Cut to fit to protect bony prominences and sensitive areas. Contains mineral oil to help soften and protect delicate skin. Washable with soap and water. Latex-free, 4" x 4".