Learn More
Research Products International Corp Clear Tip Open Bin, 5 Units, 5 1/4 x 23 5/8 x 6 1/2 Inches
SDP
Catalog No. 50212744
Encompass
Convenient bins provide an efficient and orderly solution for storing common laboratory items. Clear bins allow full visibility of contents and can be completely removed for cleaning. Bins tip open individually to a 45° angle for complete access to stored items. Heavy duty plastic housing can be stacked or wall mounted. Attractive ivory color.
Catalog No. 50-212-744 Supplier Research Products International Corp Supplier No. 560201
Convenient bins provide an efficient and orderly solution for storing common laboratory items. Clear bins allow full visibility of contents and can be completely removed for cleaning. Bins tip open individually to a 45° angle for complete access to stored items. Heavy duty plastic housing can be stacked or wall mounted. Attractive ivory color.