Learn More
Genesee Scientific Corporation CLEAR TUBES 1.7ML 500/PK
SDP
Catalog No. NC9420237
Encompass
Axygen microcentrifuge tubes are engineered to close with a "snap" and stay closed during boiling and centrifugation. They have been tested to 55 psi and are ready for autoclaving at 121�C for 15 minutes. The tubes are exceptionally clear with frosted caps for labeling, a 0.28mm piercing port, and can be used in any centrifuge up to 20,000 x g. Boilproof, Polypropylene, Box of 500 Tubes/Unit. Exceptionally clear - glass-like clarity.
Catalog No. NC9420237 Supplier Genesee Scientific Corporation Supplier No. 22282
Axygen microcentrifuge tubes are engineered to close with a "snap" and stay closed during boiling and centrifugation. They have been tested to 55 psi and are ready for autoclaving at 121�C for 15 minutes. The tubes are exceptionally clear with frosted caps for labeling, a 0.28mm piercing port, and can be used in any centrifuge up to 20,000 x g. Boilproof, Polypropylene, Box of 500 Tubes/Unit. Exceptionally clear - glass-like clarity.