Learn More
Andwin Scientific CLEAR TUBES 1.7ML
SDP
Catalog No. NC9580130
Encompass_Preferred
The GeneMate 1.7 mL Graduated Microcentrifuge Tubes are ideal for boiling procedures. The tubes have an unmatched strength and clarity with its virgin polypropylene material. This product features flat cap that will not pop open during boiling, but is still easy to open and close. The cap also has a frosted writing surface. The tubes have a volume capacity of 1.7 mL.
Catalog No. NC9580130 Supplier Andwin Scientific Supplier No. C32601
The GeneMate 1.7 mL Graduated Microcentrifuge Tubes are ideal for boiling procedures. The tubes have an unmatched strength and clarity with its virgin polypropylene material. This product features flat cap that will not pop open during boiling, but is still easy to open and close. The cap also has a frosted writing surface. The tubes have a volume capacity of 1.7 mL.