Learn More
Bellco Glass, Inc. Clear Vial,2.0mL - 8-425 Thd 12x32mm
SDP
Catalog No. 501945354
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed to work in a variety of auto-samplers requiring narrow neck vials. Narrow opening allows use of 8-425 closures and septa. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmVial Volume: 2.0mlCap Size: 8-425mm Not IncludedCase Quantity: 1000
Catalog No. 50-194-5354 Supplier Bellco Glass, Inc. Supplier No. 207000010
Manufactured from Type 1 Class A Borosilicate Glass. Designed to work in a variety of auto-samplers requiring narrow neck vials. Narrow opening allows use of 8-425 closures and septa. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmVial Volume: 2.0mlCap Size: 8-425mm Not IncludedCase Quantity: 1000