Learn More
Bellco Glass, Inc. Clear Vial w/Cap,2.0mL
SDP
Supplier:  Bellco Glass, Inc. 207000210
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Amber vials are manufactures from type 1 amber glass. these vials fit most auto-sampler. Designed for use with varian and other auto-samplers. MADE IN THE USA. Size: 12x32mm Amber & clear vials available
Catalog No. 50-194-5376