Learn More
Bellco Glass, Inc. Clear Vial W/Grad Line,2.0mL 11mm Crimp Top
SDP
Catalog No. 501945367
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Amber vials are manufactured from type 1 amber glass. Vials accept 11mm aluminum seals. Large opening vials have wider id for easier sample access. Designed for use with Hewlett Packard and other autosamplers. MADE IN THE USA. Dimensions: 12mm OD x 32mm HeightVial Volume: 2.0mLVial Material: Clear type 1 borosilicate glassCap Size: 11mmShelf Pack: 100Case Pack: 1000
Catalog No. 50-194-5367 Supplier Bellco Glass, Inc. Supplier No. 207000130
Manufactured from type 1 class a Borosilicate glass. Amber vials are manufactured from type 1 amber glass. Vials accept 11mm aluminum seals. Large opening vials have wider id for easier sample access. Designed for use with Hewlett Packard and other autosamplers. MADE IN THE USA. Dimensions: 12mm OD x 32mm HeightVial Volume: 2.0mLVial Material: Clear type 1 borosilicate glassCap Size: 11mmShelf Pack: 100Case Pack: 1000