Learn More
Chemglass Life Sciences 12mm x 32mm Clear Vial, 2ml, Graduated, 10-425 Threaded Top
SDP
Catalog No. 501531774
Encompass_Preferred
12mm x 32mm Clear Vial, 2ml, Graduated, 10-425 Threaded Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 12mm Dia. x 32mm long threaded vials comes complete with the corresponding stir bar already inside each vial. Threaded closures are also supplied in separate bag. The components are packaged in clear trays with clear snap on lids. The trays fit easily into bench drawers for easy access and the packaging keeps products visible & particle free.
Catalog No. 50-153-1774 Supplier Chemglass Life Sciences Supplier No. CG4901125
12mm x 32mm Clear Vial, 2ml, Graduated, 10-425 Threaded Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 12mm Dia. x 32mm long threaded vials comes complete with the corresponding stir bar already inside each vial. Threaded closures are also supplied in separate bag. The components are packaged in clear trays with clear snap on lids. The trays fit easily into bench drawers for easy access and the packaging keeps products visible & particle free.