Learn More
Chemglass Life Sciences 12mm x 32mm Clear Vial, 2ml, Graduated, Silver Aluminum Crimp Top
SDP
Catalog No. 501531773
Encompass_Preferred
12mm x 32mm Clear Vial, 2ml, Graduated, Silver Aluminum Crimp Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 12mm Dia. x 32mm long crimp top vials comes complete with the corresponding stir bar already inside each vial. Crimp top closures & Septa are also supplied in separate bag. The components are packaged in clear trays with clear snap on lids. The trays fit easily into bench drawers for easy access and the packaging keeps products visible & particle free.
Catalog No. 50-153-1773 Supplier Chemglass Life Sciences Supplier No. CG4901120
12mm x 32mm Clear Vial, 2ml, Graduated, Silver Aluminum Crimp Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 12mm Dia. x 32mm long crimp top vials comes complete with the corresponding stir bar already inside each vial. Crimp top closures & Septa are also supplied in separate bag. The components are packaged in clear trays with clear snap on lids. The trays fit easily into bench drawers for easy access and the packaging keeps products visible & particle free.