Learn More
Chemglass Life Sciences 15mm x 45mm Clear Vial, 4ml, 13-425 Threaded Top
SDP
Catalog No. 501531775
Encompass_Preferred
15mm x 45mm Clear Vial, 4ml, 13-425 Threaded Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 15mm Dia. x 45mm long threaded vials comes complete with the corresponding stir bar already inside each vial. Threaded closures are also supplied in separate bag. Vials are packaged in convenient LAB-PACS of 144 pieces.
Catalog No. 50-153-1775 Supplier Chemglass Life Sciences Supplier No. CG4901150
15mm x 45mm Clear Vial, 4ml, 13-425 Threaded Top. Chemglass has made it easier for you by providing convenient pre-packed vials. Every pack of 15mm Dia. x 45mm long threaded vials comes complete with the corresponding stir bar already inside each vial. Threaded closures are also supplied in separate bag. Vials are packaged in convenient LAB-PACS of 144 pieces.