Learn More
Uline Clearview Cabinet 36 x 24 x 72", Assembled
Catalog No. NC2574882
Encompass
Tan Freestanding industrial-grade steel storage cabinet, clear acrylic panels, lockable door, adjustable shelves, reinforced steel doors
Catalog No. NC2574882 Supplier Uline Supplier No. H3109AT
Tan Freestanding industrial-grade steel storage cabinet, clear acrylic panels, lockable door, adjustable shelves, reinforced steel doors