Learn More
MSC Quantum Storage Systems Hulk Container, Polypropylene, 24 in., 1.52 cu. ft., 4/Pk.
Catalog No. 5000514228
Encompass
The Clear-View container is a large, heavy-duty injection-molded bin designed for efficient storage and organization. Constructed from durable polypropylene, this container features a transparent design for easy content identification and is suitable for a wide range of industrial applications.
  • Stackable design with a wide ledge and anti-slide locks to prevent shifting
  • Autoclavable up to 250°F (121°C)
  • Resistant to extreme cold, rust, and corrosion
  • Heavy-duty grips on the front, back, and sides for easy handling
  • Includes label holder
  • Watertight construction
  • Optional dividers and clear plastic windows available
Catalog No. 50-005-14228 Supplier MSC Supplier No. 58662990
The Clear-View container is a large, heavy-duty injection-molded bin designed for efficient storage and organization. Constructed from durable polypropylene, this container features a transparent design for easy content identification and is suitable for a wide range of industrial applications.
  • Stackable design with a wide ledge and anti-slide locks to prevent shifting
  • Autoclavable up to 250°F (121°C)
  • Resistant to extreme cold, rust, and corrosion
  • Heavy-duty grips on the front, back, and sides for easy handling
  • Includes label holder
  • Watertight construction
  • Optional dividers and clear plastic windows available