Learn More
MSC Quantum Storage Systems Storage Bin, Tri-Clear Polypropylene, 16.5 in. x 10.9 in. x 5 in., 6/Pk.
Catalog No. 5000514460
Encompass
The Clear-View Ultra hang and stack bin provides a durable solution for organizing inventory. These bins are constructed from clear, high-quality material, allowing for quick and easy identification of contents, and are designed to securely stack or hang from louvered panels and rails.
  • Constructed from durable tri-clear polypropylene
  • Features a built-in rear hanger for mounting on louvered panels or rails
  • Includes a stacking ledge and anti-slide lock for stable stacking
  • Open hopper front allows easy access to stored items
  • Equipped with back and side grips for convenient handling
  • Molded-in identification card slot
  • Resistant to extreme cold
  • Load capacity of 75 lb.
Catalog No. 50-005-14460 Supplier MSC Supplier No. 58662867
The Clear-View Ultra hang and stack bin provides a durable solution for organizing inventory. These bins are constructed from clear, high-quality material, allowing for quick and easy identification of contents, and are designed to securely stack or hang from louvered panels and rails.
  • Constructed from durable tri-clear polypropylene
  • Features a built-in rear hanger for mounting on louvered panels or rails
  • Includes a stacking ledge and anti-slide lock for stable stacking
  • Open hopper front allows easy access to stored items
  • Equipped with back and side grips for convenient handling
  • Molded-in identification card slot
  • Resistant to extreme cold
  • Load capacity of 75 lb.