Learn More
MSC Quantum Storage Systems Stack And Hang Bin, Polypropylene, 13.71 Qt., Clear, 4/Pk.
Catalog No. 5000511689
Encompass
These stackable and hangable bins are designed for efficient organization in workshops, garages, and business environments. Constructed from durable, FDA-approved polypropylene, these containers feature a clear design that allows for quick identification of contents.
  • Capacity of 13.71 qt. per bin
  • Durable, waterproof polypropylene construction
  • Reinforced rib design for secure stacking and stability
  • Integrated front label slot for organized content management
  • Autoclavable up to 250°F
  • Features convenient carrying handles
  • Dimensions measure 11 in. W x 16 in. D x 8 in. H
  • Package includes 4 bins
Catalog No. 50-005-11689 Supplier MSC Supplier No. 58662719
These stackable and hangable bins are designed for efficient organization in workshops, garages, and business environments. Constructed from durable, FDA-approved polypropylene, these containers feature a clear design that allows for quick identification of contents.
  • Capacity of 13.71 qt. per bin
  • Durable, waterproof polypropylene construction
  • Reinforced rib design for secure stacking and stability
  • Integrated front label slot for organized content management
  • Autoclavable up to 250°F
  • Features convenient carrying handles
  • Dimensions measure 11 in. W x 16 in. D x 8 in. H
  • Package includes 4 bins