Learn More
Sigma Aldrich Fine Chemicals Biosciences Clear-viewTM Snap-Cap micr
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T4816250EA
Encompass
Superior clarity for sample viewingA consistent ring-lock seal prevents inadvertent openingAngled cap lid makes opening easy without thumb fatigueLarge flat tops for ease of identification and a pierceable capLarge crisp reference lines and graduation numbersCertified RNase/DNase free
Catalog No. 50-290-1484