Learn More
Sigma Aldrich Fine Chemicals Biosciences Clear-viewTM Snap-Cap micr
Catalog No. 502900930
Encompass
Superior clarity for sample viewingA consistent ring-lock seal prevents inadvertent openingAngled cap lid makes opening easy without thumb fatigueLarge flat tops for ease of identification and a pierceable capLarge crisp reference lines and graduation numbersCertified RNase/DNase free
Catalog No. 50-290-0930 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T3566500EA
Superior clarity for sample viewingA consistent ring-lock seal prevents inadvertent openingAngled cap lid makes opening easy without thumb fatigueLarge flat tops for ease of identification and a pierceable capLarge crisp reference lines and graduation numbersCertified RNase/DNase free