Learn More
Carson Manufacturing Clear Vinyl Durapron Aprons, 4mil, no hem
SDP
Catalog No. NC9465381
Encompass
This item has a minimum qty of 100 per supplier requirements.
Carson Durapron Aprons safeguard against water, oil, grease, and most chemicals, are unaffected by extreme temperature changes and will not support combustion. Individually labeled and sealed in plastic bags, our aprons have electronically
This item is not returnable. View return policy
Catalog No. NC9465381 Supplier Carson Manufacturing Supplier No. APRON FS1110
This item has a minimum qty of 100 per supplier requirements.
Carson Durapron Aprons safeguard against water, oil, grease, and most chemicals, are unaffected by extreme temperature changes and will not support combustion. Individually labeled and sealed in plastic bags, our aprons have electronically