Learn More
Cole-Parmer CLEAR VOA VIALS, 20 ML, 72/CS
Supplier:  Cole-Parmer 3520493
Encompass
Environmental Express APC3675P Clear VOA Vials, 20 mL, PTFE-Lined Cap; 72/CsPerfect for general useLevel 3 vials meet EPA standards but do not contain a certificate of analysisideal for every-day applications
Catalog No. NC2266925