Learn More
GRANT INSTRUMENTS Lid gabled polycarbonate clear for SAP18/26, JBN18/26, JBA18 and SBB AQUA 26 PLUS
SDP
Catalog No. NC1628024
Encompass_Preferred
Lid gabled polycarbonate clear for SAP18/26, JBN18/26, JBA18 and SBB AQUA 26 PLUS. Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Catalog No. NC1628024 Supplier GRANT INSTRUMENTS Supplier No. AQL26
Lid gabled polycarbonate clear for SAP18/26, JBN18/26, JBA18 and SBB AQUA 26 PLUS. Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.