Learn More
Leinco Technologies Inc ClearFlow™ Sheath Fluid with Surfactant
SDP
Catalog No. NC0400360
Encompass_Preferred
ClearFlow™ Sheath Fluid with Surfactant has been developed specifically for use on flow cytometers with high- throughput. capabilities (HTS) to prevent air bubble formation. ClearFlow™ with Surfactant is an isotonic, pH 7.2 buffered saline solution manufactured and ultra-filtered to guarantee higher signal to noise ratio measurements when doing small particle analysis.
Catalog No. NC0400360 Supplier Leinco Technologies Inc Supplier No. S64420 L
ClearFlow™ Sheath Fluid with Surfactant has been developed specifically for use on flow cytometers with high- throughput. capabilities (HTS) to prevent air bubble formation. ClearFlow™ with Surfactant is an isotonic, pH 7.2 buffered saline solution manufactured and ultra-filtered to guarantee higher signal to noise ratio measurements when doing small particle analysis.