Learn More
Zymo Research Corporation Clearing Beads, 1 ml
SDP
Catalog No. 502037032
Encompass_Preferred
Component used in the Quick-DNA Urine kit (Cat #D3061). Assists in visualizing the cellular- and nucleic acid- containing  pellet after the addition of Urine Conditioning Buffer (Cat # D3061-1-8) to a urine sample.
Catalog No. 50-203-7032 Supplier Zymo Research Corporation Supplier No. D306121
Component used in the Quick-DNA Urine kit (Cat #D3061). Assists in visualizing the cellular- and nucleic acid- containing  pellet after the addition of Urine Conditioning Buffer (Cat # D3061-1-8) to a urine sample.