Learn More
MSC Krylon Omni-Fill Valve Clearing Tool, For Male Valve, 1 ea
Catalog No. 502818621
Encompass
The Krylon Omni-Fill Valve Clearing Tool helps clear the spray mechanism before seating the actuator. This device is designed to speed up and simplify the finishing process when multiple cans are being filled, and it also prevents clogging and clears the valve stem.
  • Clears the spray mechanism before seating the actuator
  • Speeds up and simplifies the finishing process for multiple cans
  • Prevents clogging and clears valve stem
Catalog No. 50-281-8621 Supplier MSC Supplier No. 37595295
The Krylon Omni-Fill Valve Clearing Tool helps clear the spray mechanism before seating the actuator. This device is designed to speed up and simplify the finishing process when multiple cans are being filled, and it also prevents clogging and clears the valve stem.
  • Clears the spray mechanism before seating the actuator
  • Speeds up and simplifies the finishing process for multiple cans
  • Prevents clogging and clears valve stem