Learn More
Electron Microscopy Sciences Clearium Mounting Media Mounting Media in Dripper Bottle 4 OZ
SDP
Catalog No. 501926794
Encompass_Preferred
Clear and coverslip your slide preparations in one easy step. This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol .
Catalog No. 50-192-6794 Supplier Electron Microscopy Sciences Supplier No. 1352013
Clear and coverslip your slide preparations in one easy step. This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol .