Learn More
Electron Microscopy Sciences Clearium Mounting Media Mounting Media 16 OZ
SDP
Catalog No. 501926793
Encompass_Preferred
Clear and coverslip your slide preparations in one easy step. This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol .
Catalog No. 50-192-6793 Supplier Electron Microscopy Sciences Supplier No. 1352012
Clear and coverslip your slide preparations in one easy step. This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol .