Learn More
Electron Microscopy Sciences Clearium Mounting Media Mounting Media 16 OZ
SDP
Supplier:  Electron Microscopy Sciences 1352011
Encompass_Preferred
Clear and coverslip your slide preparations in one easy step. This unique product eliminates the need for hazardous xylene or xylene substitutes as a clearing agent in histology and cytology laboratories. With Clearium, simply do away with the final step of clearing and coverslip directly from absolute isopropyl alcohol .
Catalog No. 50-192-6792