Learn More
StatLab ClearMount Fast-Drying Mounting Media
SDP
Catalog No. 22000067
Change view
Click to view available options
Volume (Metric):
100 mL
Catalog No. Volume (Metric)
22-000-067 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-067 Supplier StatLab Supplier No. MMC0126
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
ClearMount is one of the fastest drying mounting medium ever formulated
  • Optically clear as glass and completely neutral
  • Compatible with virtually all clearing agents
  • Formulated for manual or automated coverslipping
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 100 mL