Learn More
StatLab ClearMount Mounting Medium
SDP
Catalog No. 22021429
Change view
Click to view available options
Volume (Metric):
500 mL
Catalog No. Volume (Metric)
22-021-429 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-429 Supplier StatLab Supplier No. MMC0143
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab ClearMount Mounting Medium - a fast-drying, permanent mounting medium
  • One of the fastest drying mounting medium ever formulated
  • It is optically clear as glass and completely neutral
  • Formula is compatible with virtually all clearing agents
  • ClearMount is formulated for manual or automated coverslipping
Specifications
Product Type ClearMount Mounting Solution
Reagent Type Mounting Solution
Volume (Metric) 500 mL