Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDSOURCE INTERNATIONAL LLC ClearSafe (PTFE) IV CATHETER 22g x 1"

Supplier:  MEDSOURCE INTERNATIONAL LLC MS84122

Encompass_Preferred

ClearSafe (PTFE) IV CATHETER 22g x 1"

Catalog No. NC2698925


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.